Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127355.1 | NP_001120827.1 | 4 - 5 | 678 | 148 |
NM_001127356.1 | NP_001120828.1 | 3 - 4 | 900 | 148 | |
NM_010122.2 | NP_034252.2 | 4 - 5 | 533 | 148 | |
GenBank mRNA | AK154364.1 | - | 4 - 5 | 441 | 147 |
AK158282.1 | - | 4 - 5 | 442 | 148 | |
BC021884.1 | - | 4 - 5 | 420 | 148 | |
M98035.1 | - | 3 - 4 | 520 | 148 | |
M98036.1 | - | 4 - 5 | 486 | 148 |