Assay Design
Gene Transcripts
- Gene Symbol
- Fga
- Entrez Gene ID
-
14161
- Gene Name
- fibrinogen alpha chain
- Gene Aliases
-
ENSMUSG00000059807, Fib
- Chromosome Location
-
Chr.3: 83026153 - 83033627 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK146828.1 |
- |
1 - 2 |
107 |
148 |
AW105859.1 |
- |
1 - 2 |
109 |
148 |
BX525925.1 |
- |
1 - 2 |
134 |
148 |
View Full Product Details