Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172689.3 | NP_766277.3 | 2 - 3 | 365 | 75 |
GenBank mRNA | AK052871.1 | - | 2 - 3 | 263 | 75 |
AK078287.1 | - | 2 - 3 | 297 | 75 | |
AK087261.1 | - | 2 - 3 | 366 | 75 | |
AK128929.1 | - | 2 - 3 | 300 | 75 | |
AK155930.1 | - | 2 - 3 | 301 | 75 | |
AY553221.1 | - | 2 - 3 | 241 | 75 | |
BC027369.1 | - | 2 - 3 | 344 | 75 | |
BC106095.1 | - | 2 - 3 | 285 | 75 | |
BC127163.1 | - | 2 - 3 | 231 | 75 | |
BC152540.1 | - | 2 - 3 | 233 | 75 |