Assay Design



Gene Transcripts

Gene Symbol
Evi5l
Entrez Gene ID
213027
Gene Name
ecotropic viral integration site 5 like
Gene Aliases
1700084G18Rik, 2310039H16Rik, 3110007G05Rik, B130050I23Rik
Chromosome Location
Chr.8: 4166538 - 4208050 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039578.3 NP_001034667.3 1 - 2 195 76
GenBank mRNA AK045240.1 - 1 - 2 196 76
AK087482.1 - 1 - 2 196 76
BC042653.1 - 1 - 2 175 76

View Full Product Details