Assay Design



Gene Transcripts

Gene Symbol
Lamp1
Entrez Gene ID
16783
Gene Name
lysosomal-associated membrane protein 1
Gene Aliases
AI196048, CD107a, Lamp-1
Chromosome Location
Chr.8: 13159134 - 13175338 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010684.2 NP_034814.2 2 - 3 423 85
GenBank mRNA AK004637.1 - 2 - 3 290 85
AK159096.1 - 2 - 3 289 85
AK159376.1 - 2 - 3 262 85
AK172007.1 - 2 - 3 305 85
AK187750.1 - 1 - 2 69 85
AK212961.1 - 1 - 2 69 85
AK213434.1 - 1 - 2 69 85
AK214981.1 - 1 - 2 69 85
AY069968.1 - 2 - 3 360 85
BC049097.1 - 2 - 3 397 85
J03881.1 - 1 - 2 97 85
M25244.1 - 2 - 3 278 85
M32015.1 - 2 - 3 247 85

View Full Product Details