Assay Design



Gene Transcripts

Gene Symbol
Rad54b
Entrez Gene ID
623474
Gene Name
RAD54 homolog B (S. cerevisiae)
Gene Aliases
E130016E03Rik
Chromosome Location
Chr.4: 11558920 - 11615808 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256145.1 NP_001243074.1 3 - 4 360 66
GenBank mRNA AK078439.1 - 3 - 4 338 66

View Full Product Details