Assay Design



Gene Transcripts

Gene Symbol
Rad54b
Entrez Gene ID
623474
Gene Name
RAD54 homolog B (S. cerevisiae)
Gene Aliases
E130016E03Rik
Chromosome Location
Chr.4: 11558920 - 11615808 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039556.3 NP_001034645.1 11 - 12 2031 120
GenBank mRNA BC057604.1 - 11 - 12 1979 120

View Full Product Details