Assay Design
Gene Transcripts
- Gene Symbol
- H2-K2
- Entrez Gene ID
-
630499
- Gene Name
- histocompatibility 2, K region locus 2
- Gene Aliases
-
EG630499, Gm7035, H-2K1, H-2K1K, H-2K2, H2-K1
- Chromosome Location
-
Chr.17: 33974659 - 33978791 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_004446.1 |
|
3 - 4 |
508 |
64 |
GenBank mRNA |
AV447662.1 |
- |
1 - 2 |
46 |
64 |
BI111965.1 |
- |
1 - 2 |
304 |
64 |
View Full Product Details