Assay Design
Gene Transcripts
- Gene Symbol
- Setd4
- Entrez Gene ID
-
224440
- Gene Name
- SET domain containing 4
- Gene Aliases
-
AY037804, C21orf18, ORF21
- Chromosome Location
-
Chr.16: 93583457 - 93604833 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC029101.1 |
- |
1 - 2 |
160 |
69 |
View Full Product Details