Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144541.1 | NP_653124.1 | 6 - 7 | 620 | 119 |
NM_181279.1 | NP_851796.1 | 8 - 9 | 731 | 119 | |
NM_181280.1 | NP_851797.1 | 7 - 8 | 629 | 119 | |
NM_181281.1 | NP_851798.1 | 5 - 6 | 464 | 119 | |
NM_181282.1 | NP_851799.1 | 6 - 7 | 541 | 119 | |
GenBank mRNA | AF440752.1 | - | 6 - 7 | 620 | 119 |
AF527952.1 | - | 8 - 9 | 731 | 119 | |
AF527953.1 | - | 6 - 7 | 541 | 119 | |
AF527954.1 | - | 6 - 7 | 551 | 119 | |
AF527955.1 | - | 5 - 6 | 464 | 119 | |
AF538925.1 | - | 7 - 8 | 629 | 119 | |
AK050695.1 | - | 6 - 7 | 665 | 119 | |
AK080991.1 | - | 6 - 7 | 656 | 119 | |
AK156929.1 | - | 6 - 7 | 712 | 119 | |
AK192409.1 | - | 2 - 3 | 120 | ||
BC061000.1 | - | 6 - 7 | 696 | 119 | |
BC100565.1 | - | 2 - 3 | 175 | 119 |