Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030127.3 | NP_084403.2 | 6 - 7 | 1500 | 63 |
GenBank mRNA | AK035194.1 | - | 6 - 7 | 1503 | 63 |
AY037300.1 | - | 6 - 7 | 1285 | 63 | |
AY156509.1 | - | 6 - 7 | 1192 | 63 | |
BC138587.1 | - | 6 - 7 | 1224 | 63 | |
BC138588.1 | - | 6 - 7 | 1224 | 63 |