Assay Design



Gene Transcripts

Gene Symbol
Ddhd2
Entrez Gene ID
72108
Gene Name
DDHD domain containing 2
Gene Aliases
2010305K11Rik, SAMWD1, mKIAA0725
Chromosome Location
Chr.8: 25725324 - 25754590 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028102.1 NP_082378.1 9 - 10 1229 122
GenBank mRNA BC046229.1 - 8 - 9 1149 122
BC118962.2 - 9 - 10 1465 122

View Full Product Details