Assay Design



Gene Transcripts

Gene Symbol
Plekha2
Entrez Gene ID
83436
Gene Name
pleckstrin homology domain-containing, family A (phosphoinositide binding specific) member 2
Gene Aliases
6430512N22Rik, AI851530, AU041791, TAPP2, W91773
Chromosome Location
Chr.8: 25039144 - 25104917 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031257.3 NP_112547.1 8 - 9 874 85
GenBank mRNA AF286161.1 - 8 - 9 828 85
AF418551.1 - 7 - 8 740 85
AK032325.1 - 8 - 9 883 85
AK042324.1 - 8 - 9 848 85
AK078212.1 - 8 - 9 853 85
AK088395.1 - 8 - 9 816 85
AK153117.1 - 8 - 9 846 85
BC010215.1 - 8 - 9 795 85

View Full Product Details