Assay Design
Gene Transcripts
- Gene Symbol
- Mx1
- Entrez Gene ID
-
17857
- Gene Name
- MX dynamin-like GTPase 1
- Gene Aliases
-
AI893580, Mx, Mx-1
- Chromosome Location
-
Chr.16: 97447035 - 97462906 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK076430.1 |
- |
5 - 6 |
442 |
164 |
View Full Product Details