Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136236.1 | NP_001129708.1 | 8 - 9 | 935 | 92 |
NM_153090.2 | NP_694730.2 | 10 - 11 | 1124 | 92 | |
NM_178165.4 | NP_835459.1 | 9 - 10 | 1064 | 92 | |
GenBank mRNA | AF329485.1 | - | 9 - 10 | 991 | 92 |
AK036445.1 | - | 4 - 5 | 1300 | 92 | |
AK038494.1 | - | 9 - 10 | 1004 | 92 | |
AK041750.1 | - | 10 - 11 | 1125 | 92 | |
AK156528.1 | - | 9 - 10 | 1005 | 92 | |
AK156985.1 | - | 8 - 9 | 875 | 92 | |
AY158088.1 | - | 9 - 10 | 984 | 92 | |
AY506554.1 | - | 9 - 10 | 997 | 92 | |
AY506555.1 | - | 8 - 9 | 868 | 92 |