Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029626.2 | NP_083902.2 | 2 - 3 | 618 | 100 |
GenBank mRNA | AK010388.1 | - | 2 - 3 | 347 | 100 |
BC055709.1 | - | 2 - 3 | 375 | 100 | |
BC058731.1 | - | 2 - 3 | 375 | 100 | |
BC069873.1 | - | 2 - 3 | 168 | 100 |