Assay Design



Gene Transcripts

Gene Symbol
Mtx1
Entrez Gene ID
17827
Gene Name
metaxin 1
Gene Aliases
Gcap6, Mtx
Chromosome Location
Chr.3: 89209081 - 89214347 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161824.1 NP_001155296.1 3 - 4 668 68

View Full Product Details