Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025448.3 | NP_079724.1 | 1 - 2 | 289 | |
GenBank mRNA | AK005340.1 | - | 2 - 3 | 289 | |
AK010565.1 | - | 2 - 3 | 289 | ||
AK050505.1 | - | 1 - 2 | 289 | ||
AK084530.1 | - | 2 - 3 | 289 | ||
BC010214.1 | - | 1 - 2 | 289 |