Assay Design



Gene Transcripts

Gene Symbol
Wdr95
Entrez Gene ID
381693
Gene Name
WD40 repeat domain 95
Gene Aliases
4930434E21Rik
Chromosome Location
Chr.5: 149528679 - 149611894 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029440.3 NP_083716.2 14 - 15 1201 123
GenBank mRNA AK015309.1 - 12 - 13 1305 123
AK087389.1 - 14 - 15 1201 123

View Full Product Details