Assay Design



Gene Transcripts

Gene Symbol
Wdr95
Entrez Gene ID
381693
Gene Name
WD40 repeat domain 95
Gene Aliases
4930434E21Rik
Chromosome Location
Chr.5: 149528679 - 149611894 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029440.3 NP_083716.2 19 - 20 1701 72
GenBank mRNA AK015309.1 - 17 - 18 1805 72
AK087389.1 - 19 - 20 1701 72

View Full Product Details