Assay Design
Gene Transcripts
- Gene Symbol
- Wdr95
- Entrez Gene ID
-
381693
- Gene Name
- WD40 repeat domain 95
- Gene Aliases
-
4930434E21Rik
- Chromosome Location
-
Chr.5: 149528679 - 149611894 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_029440.3 |
NP_083716.2 |
6 - 7 |
488 |
93 |
GenBank mRNA |
AK087389.1 |
- |
6 - 7 |
488 |
93 |
View Full Product Details