Assay Design



Gene Transcripts

Gene Symbol
Brca2
Entrez Gene ID
12190
Gene Name
breast cancer 2
Gene Aliases
Fancd1, RAB163
Chromosome Location
Chr.5: 150522021 - 150570147 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081001.2 NP_001074470.1 25 - 26 9755 88
NM_009765.3 NP_033895.2 25 - 26 9561 88
GenBank mRNA AK132864.1 - 2 - 3 338 88
BC059904.1 - 15 - 16 3018 88
U82270.1 - 24 - 25 9247 88
U89652.1 - 24 - 25 9285 88

View Full Product Details