Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013378.2 | NP_001013396.2 | 3 - 4 | 544 | 73 |
NM_001115149.1 | NP_001108621.1 | 3 - 4 | 544 | 73 | |
NM_001115150.1 | NP_001108622.1 | 2 - 3 | 375 | 73 | |
NM_001115151.1 | NP_001108623.1 | 2 - 3 | 375 | 73 | |
NM_001115153.1 | NP_001108625.1 | 4 - 5 | 721 | 73 | |
NM_001286824.1 | NP_001273753.1 | 3 - 4 | 502 | 73 | |
GenBank mRNA | AK050969.1 | - | 3 - 4 | 528 | 73 |
AK088787.1 | - | 2 - 3 | 241 | 73 | |
AK161654.1 | - | 2 - 3 | 222 | 73 | |
AK168981.1 | - | 3 - 4 | 396 | 73 | |
BB655282.1 | - | 3 - 4 | 523 | 73 | |
BC028849.1 | - | 3 - 4 | 438 | 73 | |
BC086664.1 | - | 3 - 4 | 386 | 73 | |
BY298649.1 | - | 2 - 3 | 272 | 73 |