Assay Design



Gene Transcripts

Gene Symbol
Mapre3
Entrez Gene ID
100732
Gene Name
microtubule-associated protein, RP/EB family, member 3
Gene Aliases
AI790651, EB2, EB3
Chromosome Location
Chr.5: 30814614 - 30866106 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133350.1 NP_579928.1 1 - 2 143 75
GenBank mRNA AF468005.1 - 1 - 2 143 75
BC048833.1 - 1 - 2 145 75
BC057918.1 - 1 - 2 143 75

View Full Product Details