Assay Design



Gene Transcripts

Gene Symbol
Dcst1
Entrez Gene ID
77772
Gene Name
DC-STAMP domain containing 1
Gene Aliases
A330106H01Rik
Chromosome Location
Chr.3: 89350235 - 89365239 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK030269.1 - 14 - 15 1940 104
BC125478.1 - 14 - 15 1925 104
BC125480.1 - 14 - 15 1925 104

View Full Product Details