Assay Design



Gene Transcripts

Gene Symbol
Dcst1
Entrez Gene ID
77772
Gene Name
DC-STAMP domain containing 1
Gene Aliases
A330106H01Rik
Chromosome Location
Chr.3: 89350235 - 89365239 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK030269.1 - 5 - 6 714 80
AK041560.1 - 4 - 5 402 80
BC125478.1 - 5 - 6 699 80
BC125480.1 - 5 - 6 699 80

View Full Product Details