Assay Design



Gene Transcripts

Gene Symbol
Pglyrp3
Entrez Gene ID
242100
Gene Name
peptidoglycan recognition protein 3
Gene Aliases
Gm420
Chromosome Location
Chr.3: 92012477 - 92032672 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207247.4 NP_997130.2 1 - 2 117 67
GenBank mRNA BB619146.1 - 2 - 3 290 67
BC128291.1 - 1 - 2 88 67

View Full Product Details