Assay Design



Gene Transcripts

Gene Symbol
Pglyrp3
Entrez Gene ID
242100
Gene Name
peptidoglycan recognition protein 3
Gene Aliases
Gm420
Chromosome Location
Chr.3: 92012477 - 92032672 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207247.4 NP_997130.2 6 - 7 906 117
GenBank mRNA AY518698.1 - 6 - 7 877 117
BC128291.1 - 6 - 7 877 117

View Full Product Details