Assay Design
Gene Transcripts
- Gene Symbol
- Chd7
- Entrez Gene ID
-
320790
- Gene Name
- chromodomain helicase DNA binding protein 7
- Gene Aliases
-
A730019I05Rik, Cycn, Cyn, Dz, Edy, Flo, GENA 47, GENA 60, Gena 52, Lda, Mt, Obt, Todo, WBE1, Whi, metis
- Chromosome Location
-
Chr.4: 8690406 - 8868449 on Build GRCm38
View Full Product Details