Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173047.3 | NP_766635.1 | 1 - 2 | 401 | 81 |
GenBank mRNA | AK003232.1 | - | 1 - 2 | 406 | 81 |
AK028150.1 | - | 1 - 2 | 377 | 81 | |
BC028763.1 | - | 1 - 2 | 411 | 81 | |
BC087735.1 | - | 1 - 2 | 362 | 81 | |
BC096658.1 | - | 1 - 2 | 371 | 81 |