Assay Design



Gene Transcripts

Gene Symbol
Slc27a3
Entrez Gene ID
26568
Gene Name
solute carrier family 27 (fatty acid transporter), member 3
Gene Aliases
Acsvl3, FATP3, Vlcs-3
Chromosome Location
Chr.3: 90385228 - 90390042 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011988.2 NP_036118.2 4 - 5 1246 60
GenBank mRNA AK037042.1 - 4 - 5 1006 60
AK076014.1 - 4 - 5 978 60
AK131156.1 - 2 - 3 2023 60
AK135190.1 - 4 - 5 1223 60
BC060052.1 - 4 - 5 1254 60
BC141134.1 - 4 - 5 1211 60

View Full Product Details