Assay Design



Gene Transcripts

Gene Symbol
Slc27a3
Entrez Gene ID
26568
Gene Name
solute carrier family 27 (fatty acid transporter), member 3
Gene Aliases
Acsvl3, FATP3, Vlcs-3
Chromosome Location
Chr.3: 90385228 - 90390042 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011988.2 NP_036118.2 6 - 7 1539 123
GenBank mRNA AF072758.1 - 6 - 7 1243 123
AJ577573.1 - 6 - 7 1539 123
AK037042.1 - 6 - 7 1299 123
AK076014.1 - 6 - 7 1271 123
AK135190.1 - 6 - 7 1516 123
BC141134.1 - 6 - 7 1504 123

View Full Product Details