Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172676.2 | NP_766264.2 | 2 - 3 | 405 | 76 |
GenBank mRNA | AK053048.1 | - | 2 - 3 | 359 | 76 |
AK171151.1 | - | 2 - 3 | 411 | 76 | |
BC019475.1 | - | 2 - 3 | 415 | 76 | |
BC052921.1 | - | 2 - 3 | 405 | 76 |