Assay Design



Gene Transcripts

Gene Symbol
Samd10
Entrez Gene ID
229011
Gene Name
sterile alpha motif domain containing 10
Gene Aliases
D930033N23Rik, E130318E08
Chromosome Location
Chr.2: 181595218 - 181599491 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172676.2 NP_766264.2 3 - 4 575 75
GenBank mRNA AK053048.1 - 3 - 4 529 75
AK164204.1 - 3 - 4 614 75
AK171151.1 - 3 - 4 581 75
BC019475.1 - 3 - 4 585 75
BC052921.1 - 3 - 4 575 75

View Full Product Details