Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001048192.2 | NP_001041657.1 | 5 - 6 | 758 | 90 |
NM_001294262.1 | NP_001281191.1 | 4 - 5 | 786 | 90 | |
NM_174849.3 | NP_777274.1 | 4 - 5 | 675 | 90 | |
NR_122116.1 | 4 - 5 | 742 | 90 | ||
NR_122117.1 | 4 - 5 | 742 | 90 | ||
GenBank mRNA | AK029676.1 | - | 4 - 5 | 676 | 90 |
AK034894.1 | - | 4 - 5 | 764 | 90 | |
AK035680.1 | - | 2 - 3 | 243 | 90 | |
AK044195.1 | - | 3 - 4 | 2080 | 90 | |
AK083656.1 | - | 4 - 5 | 624 | 90 | |
BC057349.1 | - | 4 - 5 | 676 | 90 | |
DQ867034.1 | - | 4 - 5 | 671 | 90 | |
DQ867035.1 | - | 4 - 5 | 671 | 90 | |
DQ867036.1 | - | 5 - 6 | 758 | 90 | |
DQ867037.1 | - | 4 - 5 | 671 | 90 | |
DQ867038.1 | - | 4 - 5 | 671 | 90 |