Assay Design
Gene Transcripts
- Gene Symbol
- Abhd13
- Entrez Gene ID
-
68904
- Gene Name
- abhydrolase domain containing 13
- Gene Aliases
-
1110065L07Rik, AI463703, AI788994
- Chromosome Location
-
Chr.8: 9977717 - 9992155 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_026868.1 |
NP_081144.1 |
2 - 3 |
407 |
87 |
GenBank mRNA |
DV655120.1 |
- |
2 - 3 |
160 |
87 |
View Full Product Details