Assay Design
Gene Transcripts
- Gene Symbol
- Tbl1x
- Entrez Gene ID
-
21372
- Gene Name
- transducin (beta)-like 1 X-linked
- Gene Aliases
-
5330429M20, Tbl1
- Chromosome Location
-
Chr.X: 77510547 - 77662985 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK036996.1 |
- |
1 - 2 |
51 |
104 |
View Full Product Details