Assay Design



Gene Transcripts

Gene Symbol
Kdr
Entrez Gene ID
16542
Gene Name
kinase insert domain protein receptor
Gene Aliases
6130401C07, Flk-1, Flk1, Krd-1, Ly73, VEGFR-2, VEGFR2, sVEGFR-2
Chromosome Location
Chr.5: 75932827 - 75979074 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010612.2 NP_034742.2 8 - 9 1361 106
GenBank mRNA AK054510.1 - 8 - 9 1394 105
AK141938.1 - 8 - 9 1384 106
BC020530.1 - 8 - 9 1361 106
EU884114.1 - 8 - 9 1391 106
S53103.1 - 8 - 9 1372 106
X59397.1 - 8 - 9 1308 106
X70842.1 - 8 - 9 1386 106

View Full Product Details