Assay Design



Gene Transcripts

Gene Symbol
Prss48
Entrez Gene ID
368202
Gene Name
protease, serine 48
Gene Aliases
Esspl, Gm1019
Chromosome Location
Chr.3: 85973843 - 86006002 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001650.1 NP_001001650.1 4 - 5 513 89
GenBank mRNA BC116378.1 - 4 - 5 513 89
BC116379.1 - 4 - 5 513 89

View Full Product Details