Assay Design



Gene Transcripts

Gene Symbol
Bcan
Entrez Gene ID
12032
Gene Name
brevican
Gene Aliases
Cspg7
Chromosome Location
Chr.3: 87987531 - 88000893 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007529.2 NP_031555.2 12 - 13 2643 88
GenBank mRNA AK144405.1 - 12 - 13 2444 88
AK163646.1 - 5 - 6 704 88
BC052032.1 - 12 - 13 2438 88
X87096.1 - 12 - 13 2519 88

View Full Product Details