Assay Design



Gene Transcripts

Gene Symbol
Gpatch4
Entrez Gene ID
66614
Gene Name
G patch domain containing 4
Gene Aliases
2610029K21Rik, Gpatc4
Chromosome Location
Chr.3: 88043106 - 88055994 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110809.2 NP_001104279.1 6 - 7 553 72
NM_001286857.1 NP_001273786.1 6 - 7 1167 72
NM_025663.4 NP_079939.3 7 - 8 688 72
GenBank mRNA AK011617.1 - 6 - 7 552 72
AK017310.1 - 6 - 7 503 72
AK018306.1 - 6 - 7 527 72
AK169111.1 - 6 - 7 542 72
BC002265.1 - 6 - 7 524 72
BC019490.1 - 7 - 8 670 72
BC080676.1 - 6 - 7 1145 72

View Full Product Details