Assay Design



Gene Transcripts

Gene Symbol
Serpini1
Entrez Gene ID
20713
Gene Name
serine (or cysteine) peptidase inhibitor, clade I, member 1
Gene Aliases
AI837402, Ns, PI-12, PI12, Spi17
Chromosome Location
Chr.3: 75557503 - 75642523 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009250.2 NP_033276.1 6 - 7 1101 138
GenBank mRNA AJ001700.1 - 6 - 7 1070 138
AK032152.1 - 6 - 7 1090 138
AK133770.1 - 6 - 7 1090 138
BC006776.1 - 6 - 7 1066 138

View Full Product Details