Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009250.2 | NP_033276.1 | 8 - 9 | 1286 | 116 |
GenBank mRNA | AJ001700.1 | - | 8 - 9 | 1255 | 116 |
AK032152.1 | - | 8 - 9 | 1275 | 116 | |
AK133770.1 | - | 8 - 9 | 1276 | 116 | |
BC006776.1 | - | 8 - 9 | 1251 | 116 |