Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252503.2 | NP_001239432.1 | 4 - 5 | 2486 | 70 |
NM_001290545.1 | NP_001277474.1 | 4 - 5 | 2486 | 70 | |
NM_181411.4 | NP_852076.1 | 4 - 5 | 2486 | 70 | |
GenBank mRNA | AK029681.1 | - | 4 - 5 | 2385 | 70 |
AK030572.1 | - | 4 - 5 | 2430 | 70 | |
AK047225.1 | - | 3 - 4 | 1550 | 70 | |
AK136815.1 | - | 3 - 4 | 1738 | 70 | |
AK138793.1 | - | 4 - 5 | 2408 | 70 | |
AK150458.1 | - | 2 - 3 | 277 | 70 | |
AK152308.1 | - | 2 - 3 | 277 | 70 | |
AK152487.1 | - | 2 - 3 | 277 | 70 | |
AK161532.1 | - | 2 - 3 | 290 | 70 | |
BC004630.1 | - | 3 - 4 | 775 | 70 | |
BC052036.1 | - | 4 - 5 | 2414 | 70 |