Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190451.1 | NP_001177380.1 | 1 - 2 | 297 | 82 |
GenBank mRNA | AK135287.1 | - | 1 - 2 | 189 | 82 |
AK145905.1 | - | 1 - 2 | 187 | 82 | |
AK161788.1 | - | 1 - 2 | 187 | 82 | |
AK165408.1 | - | 1 - 2 | 187 | 82 | |
BG088078.2 | - | 1 - 2 | 297 | 82 | |
X53929.1 | - | 1 - 2 | 214 | 82 |