Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029166.2 | NP_083442.2 | 15 - 16 | 3674 | 62 |
GenBank mRNA | AB093261.1 | - | 15 - 16 | 3639 | 62 |
AK141050.1 | - | 15 - 16 | 3683 | 62 | |
BC006745.1 | - | 2 - 3 | 248 | 62 | |
BC079645.1 | - | 7 - 8 | 2343 | 62 | |
BC132134.1 | - | 15 - 16 | 3467 | 62 | |
BC138092.1 | - | 15 - 16 | 3467 | 62 |