Assay Design
Gene Transcripts
- Gene Symbol
- Thumpd2
- Entrez Gene ID
-
72167
- Gene Name
- THUMP domain containing 2
- Gene Aliases
-
2810025A12Rik
- Chromosome Location
-
Chr.17: 81026327 - 81065127 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_028138.1 |
NP_082414.1 |
8 - 9 |
1108 |
159 |
GenBank mRNA |
AK012806.1 |
- |
8 - 9 |
1091 |
159 |
View Full Product Details