Assay Design
Gene Transcripts
- Gene Symbol
- Hs6st3
- Entrez Gene ID
-
50787
- Gene Name
- heparan sulfate 6-O-sulfotransferase 3
- Gene Aliases
-
6OST3, HS6ST-3
- Chromosome Location
-
Chr.14: 119137660 - 119872871 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_015820.3 |
NP_056635.2 |
2 - 2 |
1725 |
70 |
GenBank mRNA |
AB024567.1 |
- |
2 - 2 |
1688 |
70 |
View Full Product Details