Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011517.2 | NP_035647.2 | 6 - 7 | 574 | 130 |
GenBank mRNA | AK131949.1 | - | 6 - 7 | 498 | 130 |
BC048483.1 | - | 6 - 7 | 497 | 130 | |
BC089540.1 | - | 6 - 7 | 497 | 130 | |
BC099552.1 | - | 5 - 6 | 379 | 130 | |
BC132079.1 | - | 6 - 7 | 575 | 130 | |
BC138509.1 | - | 6 - 7 | 575 | 130 |