Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159711.1 | NP_001153183.1 | 4 - 5 | 376 | 134 |
NM_016923.2 | NP_058619.1 | 4 - 5 | 430 | 134 | |
GenBank mRNA | AB018550.1 | - | 4 - 5 | 430 | 134 |
AF168120.1 | - | 4 - 5 | 400 | 134 | |
AK019283.1 | - | 4 - 5 | 454 | 134 | |
AY641431.1 | - | 4 - 5 | 324 | 134 | |
BC116785.1 | - | 4 - 5 | 426 | 134 |